4-Amino-3-fluorobenzoic acid 98%

Supplier: Thermo Fisher Scientific
Warning

H26092.MD H26092.03
H26092.MDEA 0 EUR
H26092.MD H26092.03
4-Amino-3-fluorobenzoic acid 98%
4-Amino-3-fluorobenzoic acid

Purity: 98% CAS No.: 455-87-8 Molecular Formula: C7H6FNO2 Formula Weight: 155.13 Melting Point: 214-216° MDL No.: MFCD01660374

Formula: C₇H₆FNO₂
Melting Pt: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD01660374
CAS Number: 455-87-8
EINECS: 207-250-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR