4-Ethylphenylacetylene ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (4-Ethylphenyl)acetylene, 1-Ethyl-4-ethynylbenzene, 1-Ethynyl-4-ethylbenzene, (4-Ethylphenyl)ethyne

H26313.06
H26313.06EA 122 EUR
H26313.06
4-Ethylphenylacetylene ≥99%
4-Ethylphenylacetylene

Purity: 99% CAS No.: 40307-11-7 Molecular Formula: C10H10 Formula Weight: 130.19 Flash Point: 71°(160°F) Refractive Index: 1.5420 Boiling Point: 192° MDL No.: MFCD00173887

Formula: C₁₀H₁₀
MW: 130.19 g/mol
Boiling Pt: 192 °C
Density: 0.930
Flash Pt: 71 °C (160 °F)
Storage Temperature: Ambient
MDL Number: MFCD00173887
CAS Number: 40307-11-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR