3-Chloro-2-nitrobenzoic acid ≥97%

Supplier: Thermo Fisher Scientific
Warning

H26467.03 H26467.06
H26467.03EA 35.2 EUR
H26467.03 H26467.06
3-Chloro-2-nitrobenzoic acid ≥97%
3-Chloro-2-nitrobenzoic acid

Purity: 97% CAS No.: 4771-47-5 Molecular Formula: C7H4ClNO4 Formula Weight: 201.57 Odour: Odourless Melting Point: 234-240° Beilstein Reference: 2109828 MDL No.: MFCD00007087

Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Melting Pt: 234…240 °C
Density: 1.566 g/cm³ (18 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007087
CAS Number: 4771-47-5
EINECS: 225-313-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR