4-Benzyloxy-2-chloro-5-hydroxypyrimidine 95%

Supplier: Thermo Fisher Scientific
Warning

H26993.03
H26993.03EA 0 EUR
H26993.03
4-Benzyloxy-2-chloro-5-hydroxypyrimidine 95%
4-Benzyloxy-2-chloro-5-hydroxypyrimidine

Purity: 95% CAS No.: 885952-28-3 Molecular Formula: C11H9ClN2O2 Formula Weight: 236.66 MDL No.: MFCD06798283

Formula: C₁₁H₉ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06798283
CAS Number: 885952-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR