(-)-Bis[(S)-1-phenylethyl]amine ≥99%, ee 98%, ChiPros®

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Bis[(S)-1-phenylethyl]amine

H27106.14 H27106.06 H27106.03
H27106.14EA 624 EUR
H27106.14 H27106.06 H27106.03
(-)-Bis[(S)-1-phenylethyl]amine ≥99%, ee 98%, ChiPros®
(-)-Bis[(S)-1-phenylethyl]amine

Purity: 99% CAS No.: 56210-72-1 Molecular Formula: C16H19N Formula Weight: 225.34 Boiling Point: 98°/0.3mm MDL No.: MFCD00243087

Formula: C₁₆H₁₉N
MW: 225.34 g/mol
Boiling Pt: 98 °C (0.3 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00243087
CAS Number: 56210-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR