2-Nitroterephthalic acid 1-methyl ester ≥97%

Supplier: Thermo Fisher Scientific
Warning

H27121.06 H27121.14
H27121.06EA 24.3 EUR
H27121.06 H27121.14
2-Nitroterephthalic acid 1-methyl ester ≥97%
2-Nitroterephthalic acid 1-methyl ester

Purity: 97% CAS No.: 35092-89-8 Molecular Formula: C9H7NO6 Formula Weight: 225.17 Melting Point: 173-178° MDL No.: MFCD00024510

Formula: C₉H₇NO₆
Melting Pt: 173…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00024510
CAS Number: 35092-89-8
EINECS: 252-360-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR