5-Chloro-2-methoxyphenylboronic acid ≥97%

Supplier: Thermo Fisher Scientific
Warning

H27127.14 H27127.06
H27127.14EA 194 EUR
H27127.14 H27127.06
5-Chloro-2-methoxyphenylboronic acid ≥97%
5-Chloro-2-methoxyphenylboronic acid

Purity: 97% CAS No.: 89694-48-4 Molecular Formula: C7H8BClO3 Formula Weight: 186.40 Melting Point: 134-141° MDL No.: MFCD01318966

Formula: C₇H₈BClO₃
Melting Pt: 134…141 °C
Storage Temperature: Ambient
MDL Number: MFCD01318966
CAS Number: 89694-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR