2-Chloro-L-phenylalanine ≥98%

Supplier: Thermo Fisher Scientific

H27135.MD H27135.03
H27135.MDEA 169 EUR
H27135.MD H27135.03
2-Chloro-L-phenylalanine ≥98%
2-Chloro-L-phenylalanine

Purity: 98+% CAS No.: 103616-89-3 Molecular Formula: C9H10ClNO2 Formula Weight: 199.64 Melting Point: 196-199° MDL No.: MFCD00077921

Formula: C₉H₁₀ClNO₂
Melting Pt: 196…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00077921
CAS Number: 103616-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR