6-Bromo-5-methyl-3-pyridineboronic acid 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 6-Bromo-5-methylopyridine-3-boronic acid, 6-Bromo-5-methyl-3-pyridinylboronic acid, 2-Bromo-3-methylpyridine-5-boronic acid, (6-Bromo-5-methylpyridin-3-yl)boronic acid

H27221.03
H27221.03EA 0 EUR
H27221.03
6-Bromo-5-methyl-3-pyridineboronic acid 95%
6-Bromo-5-methyl-3-pyridineboronic acid

Purity: 95% CAS No.: 1003043-34-2 Molecular Formula: C6H7BBrNO2 Formula Weight: 215.84 MDL No.: MFCD08274468

Formula: C₆H₇BBrNO₂
MW: 215.84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08274468
CAS Number: 1003043-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR