1-Aminocyclopropanecarboxylic acid hydrochloride 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-azanylcyclopropane-1-carboxylic acid hydrochloride, ACPC, 1-aminocyclopropane-1-carboxylic acid hydrochloride, Cyclopropanecarboxylic acid, 1-amino-, hydrochloride

H27264.MD H27264.03
H27264.MDEA 73.6 EUR
H27264.MD H27264.03
1-Aminocyclopropanecarboxylic acid hydrochloride 97%
1-Aminocyclopropanecarboxylic acid hydrochloride

Purity: 97% CAS No.: 68781-13-5 Molecular Formula: C4H7NO2.HCl Formula Weight: 137.56 Melting Point: ca 220° dec. MDL No.: MFCD00012545

Formula: C₄H₈ClNO₂
MW: 137.57 g/mol
Melting Pt: ∼ 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012545
CAS Number: 68781-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR