4-Aminopyrimidine 98%

Supplier: Thermo Fisher Scientific
Warning

H27367.MD H27367.06 H27367.03
H27367.MDEA 70.1 EUR
H27367.MD H27367.06 H27367.03
4-Aminopyrimidine 98%
4-Aminopyrimidine

Purity: 98% CAS No.: 591-54-8 Molecular Formula: C4H5N3 Formula Weight: 95.11 Melting Point: 147-152° MDL No.: MFCD00006112

Formula: C₄H₅N₃
MW: 95.1 g/mol
Melting Pt: 147…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00006112
CAS Number: 591-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR