4-Iodobiphenyl ≥97%

Supplier: Thermo Fisher Scientific

H27453.14 H27453.06 H27453.03
H27453.14EA 495 EUR
H27453.14 H27453.06 H27453.03
4-Iodobiphenyl ≥97%
4-Iodobiphenyl

Purity: 97% CAS No.: 1591-31-7 Molecular Formula: C12H9I Formula Weight: 280.11 Melting Point: 110-114° UN No.: 3077 MDL No.: MFCD00019028

Formula: C₁₂H₉I
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 110…114 °C
Density: 1,584 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00019028
CAS Number: 1591-31-7
EINECS: 216-469-1
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR