4-Bromo-2-nitrobenzoic acid ≥97%

Supplier: Thermo Fisher Scientific
Warning

H27541.06 H27541.03
H27541.06EA 66.3 EUR
H27541.06 H27541.03
4-Bromo-2-nitrobenzoic acid ≥97%
4-Bromo-2-nitrobenzoic acid

Purity: 97% CAS No.: 99277-71-1 Molecular Formula: C7H4BrNO4 Formula Weight: 246.02 Melting Point: 165-169° MDL No.: MFCD01013599

Formula: C₇H₄BrNO₄
Melting Pt: 165…169 °C
Storage Temperature: Ambient
MDL Number: MFCD01013599
CAS Number: 99277-71-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR