2,6-Dichloro-p-benzoquinone ≥97%

Supplier: Thermo Fisher Scientific
Danger

H27542.14 H27542.06
H27542.14EA 385 EUR
H27542.14 H27542.06
2,6-Dichloro-p-benzoquinone ≥97%
2,6-Dichloro-p-benzoquinone

Purity: 98% CAS No.: 697-91-6 Molecular Formula: C6H2Cl2O2 Formula Weight: 176.98 Melting Point: 122-124° MDL No.: MFCD00037159

Formula: C₆H₂Cl₂O₂
Melting Pt: 119…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00037159
CAS Number: 697-91-6
EINECS: 211-810-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR