(2-Hydroxy-3-N,N,N-trimethylamino)propyl β-cyclodextrin chloride hydrate

Supplier: Thermo Fisher Scientific

Synonyms: (2-Hydroxy-3-N,N,N-trimethylamino)propyl beta-cyclodextrin chloride hydrate, Quaternary ammonium beta-cylodextrin, QA-beta-CD

H27604.MD H27604.03
H27604.MDEA 86.9 EUR
H27604.MD H27604.03
(2-Hydroxy-3-N,N,N-trimethylamino)propyl β-cyclodextrin chloride hydrate
(2-Hydroxy-3-N,N,N-trimethylamino)propyl β-cyclodextrin chloride hydrate

Melting Point: ca 280° dec. MDL No.: MFCD00078139

Melting Pt: ∼ 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00078139
EINECS: 231-493-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR