4-tert-Butyldimethylsiloxy-3-penten-2-one mixture of isomers 95%

Supplier: Thermo Fisher Scientific
Warning

H27868.06
H27868.06EA 0 EUR
H27868.06
4-tert-Butyldimethylsiloxy-3-penten-2-one mixture of isomers 95%
4-tert-Butyldimethylsiloxy-3-penten-2-one

Purity: 95% CAS No.: 69404-97-3 Molecular Formula: C11H22O2Si Formula Weight: 214.38 Flash Point: 73°(163°F) Boiling Point: 50-51°/0.2mm MDL No.: MFCD00010655

Formula: C₁₁H₂₂O₂Si
Boiling Pt: 50…51 °C (0,2 mmHg)
Density: 0,898
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010655
CAS Number: 69404-97-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR