1,2-Diphenyl-1,1,2,2-tetramethyldisilane 95%

Supplier: Thermo Fisher Scientific

H28705.06 H28705.03
H28705.06EA 152 EUR
H28705.06 H28705.03
1,2-Diphenyl-1,1,2,2-tetramethyldisilane 95%
1,2-Diphenyl-1,1,2,2-tetramethyldisilane

Purity: 97% CAS No.: 1145-98-8 Molecular Formula: C16H22Si2 Formula Weight: 270.52 MDL No.: MFCD00054786

Formula: C₁₆H₂₂Si₂
Boiling Pt: 111 °C
Melting Pt: 34…37 °C
Density: 0,97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054786
CAS Number: 1145-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR