Biphenyl diphenyl ether eutectic

Supplier: Thermo Fisher Scientific
Warning

H30347.AU H30347.0F
H30347.AUEA 137 EUR
H30347.AU H30347.0F
Biphenyl diphenyl ether eutectic
Biphenyl diphenyl ether eutectic

CAS No.: 8004-13-5 Flash Point: 113°(235°F) Odour: Unpleasant Melting Point: 12-14° Boiling Point: 257° UN No.: UN3082 MDL No.: MFCD00148859

Formula: C₂₄H₂₀O
Boiling Pt: 257 °C
Melting Pt: 12…14 °C
Density: 1,056
Flash Pt: 113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00148859
CAS Number: 8004-13-5
EINECS: 616-827-0
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR