2,5-Dibromo-4-picoline ≥97%

Supplier: Thermo Fisher Scientific
Warning

H30457.03 H30457.06
H30457.03EA 0 EUR
H30457.03 H30457.06
2,5-Dibromo-4-picoline ≥97%
2,5-Dibromo-4-picoline

Purity: 97% CAS No.: 3430-26-0 Molecular Formula: C6H5Br2N Formula Weight: 250.92 Melting Point: 36-40° MDL No.: MFCD00234955

Formula: C₆H₅Br₂N
Melting Pt: 36…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00234955
CAS Number: 3430-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR