4-Bromo-2-(trifluoromethyl)phenol 99%

Supplier: Thermo Fisher Scientific
Warning

H31787.03 H31787.06
H31787.03EA 37 EUR
H31787.03 H31787.06
4-Bromo-2-(trifluoromethyl)phenol 99%
4-Bromo-2-(trifluoromethyl)phenol

Purity: 99% CAS No.: 50824-04-9 Molecular Formula: C7H4BrF3O Formula Weight: 241.01 Melting Point: 82-86° MDL No.: MFCD05668981

Formula: C₇H₄BrF₃O
Melting Pt: 82…86 °C
Storage Temperature: Ambient
MDL Number: MFCD05668981
CAS Number: 50824-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR