3,3'-Di-n-pentylthiacarbocyanine iodide 96%

Supplier: Thermo Fisher Scientific
Warning

H31886.MEEA 0 EUR
H31886.ME
3,3'-Di-n-pentylthiacarbocyanine iodide 96%
3,3'-Di-n-pentylthiacarbocyanine iodide

Purity: 96% CAS No.: 53213-86-8 Molecular Formula: C27H33IN2S2 Formula Weight: 576.61 Melting Point: 215° dec. MDL No.: MFCD07433846

Formula: C₂₇H₃₃IN₂S₂
Melting Pt: 215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD07433846
CAS Number: 53213-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR