5-Methoxy-2-(trifluoromethyl)pyridine ≥97%

Supplier: Thermo Fisher Scientific
Warning

H32586.03 H32586.06
H32586.03EA 70.7 EUR
H32586.03 H32586.06
5-Methoxy-2-(trifluoromethyl)pyridine ≥97%
5-Methoxy-2-(trifluoromethyl)pyridine

Purity: 97% CAS No.: 216766-13-1 Molecular Formula: C7H6F3NO Formula Weight: 177.13 MDL No.: MFCD09864699

Formula: C₇H₆F₃NO
Storage Temperature: Ambient
MDL Number: MFCD09864699
CAS Number: 216766-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR