2-Fluoro-5-(trifluoromethoxy)benzamide 97%

Supplier: Thermo Fisher Scientific
Warning

H32798.09
H32798.09EA 0 EUR
H32798.09
2-Fluoro-5-(trifluoromethoxy)benzamide 97%
2-Fluoro-5-(trifluoromethoxy)benzamide

Purity: 97% CAS No.: 886498-17-5 Molecular Formula: C8H5F4NO2 Formula Weight: 223.12 MDL No.: MFCD04115896

Formula: C₈H₅F₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD04115896
CAS Number: 886498-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR