Methyl-3-aminosalicylate 97%

Supplier: Thermo Fisher Scientific
Warning

H33035.03 H33035.06
H33035.03EA 108 EUR
H33035.03 H33035.06
Methyl-3-aminosalicylate 97%
Methyl 3-aminosalicylate
Formula: C₈H₉NO₃
Melting Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD09842614
CAS Number: 35748-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR