3-Amino-5-bromo-1-methyl-1H-indazole 97%

Supplier: Thermo Fisher Scientific
Warning

H33202.MD H33202.06 H33202.03
H33202.MDEA 47 EUR
H33202.MD H33202.06 H33202.03
3-Amino-5-bromo-1-methyl-1H-indazole 97%
3-Amino-5-bromo-1-methyl-1H-indazole
Formula: C₈H₈BrN₃
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD09864804
CAS Number: 1000018-06-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR