3,5-Dichloro-1H-indazole 95%

Supplier: Thermo Fisher Scientific
Warning

H33220.03
H33220.03EA 165 EUR
H33220.03
3,5-Dichloro-1H-indazole 95%
3,5-Dichloro-1H-indazole
Formula: C₇H₄Cl₂N₂
Melting Pt: 240…242 °C
Storage Temperature: Ambient
MDL Number: MFCD09263200
CAS Number: 36760-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR