5-(2-Thienyl)isoxazole 97%

Supplier: Thermo Fisher Scientific
Warning

H33225.03
H33225.03EA 0 EUR
H33225.03
5-(2-Thienyl)isoxazole 97%
5-(2-Thienyl)isoxazole

Purity: 97% CAS No.: 138716-44-6 Molecular Formula: C7H5NOS Formula Weight: 151.19 Refractive Index: 1.6130 Boiling Point: 68-70°/0.22mm MDL No.: MFCD06409150

Formula: C₇H₅NOS
Boiling Pt: 68…70 °C (0,22 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD06409150
CAS Number: 138716-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR