5-Fluorobenzo[b]thiophene-2-carboxylic acid 96%

Supplier: Thermo Fisher Scientific
Warning

H33239.09 H33239.18 H33239.03 H33239.06
H33239.09EA 0 EUR
H33239.09 H33239.18 H33239.03 H33239.06
5-Fluorobenzo[b]thiophene-2-carboxylic acid 96%
5-Fluorobenzo[b]thiophene-2-carboxylic acid

Purity: 96% CAS No.: 70060-13-8 Molecular Formula: C9H5FO2S Formula Weight: 196.19 Melting Point: 257-262° MDL No.: MFCD01927184

Formula: C₉H₅FO₂S
Melting Pt: 257…262 °C
Storage Temperature: Ambient
MDL Number: MFCD01927184
CAS Number: 70060-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR