6-Hydroxy-m-anisidine 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Amino-4-methoxyphenol

H33724.03 H33724.06
H33724.03EA 71.4 EUR
H33724.03 H33724.06
6-Hydroxy-m-anisidine 97%
6-Hydroxy-m-anisidine
Formula: C₇H₉NO₂
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD06616911
CAS Number: 20734-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR