4-Phenoxybenzyl bromide ≥97%

Supplier: Thermo Fisher Scientific
Danger

H33895.MD H33895.03
H33895.MDEA 125 EUR
H33895.MD H33895.03
4-Phenoxybenzyl bromide ≥97%
4-Phenoxybenzyl bromide
Formula: C₁₃H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD04038693
CAS Number: 36881-42-2
EINECS: 253-253-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR