Ethyl-3-amino-1-benzothiophene-2-carboxylate 97%

Supplier: Thermo Fisher Scientific
Warning

H34050.06
H34050.06EA 0 EUR
H34050.06
Ethyl-3-amino-1-benzothiophene-2-carboxylate 97%
Ethyl-3-amino-1-benzothiophene-2-carboxylate

Purity: 97% CAS No.: 34761-09-6 Molecular Formula: C11H11NO2S Formula Weight: 221.28 Melting Point: ca 82° subl. MDL No.: MFCD00661903

Formula: C₁₁H₁₁NO₂S
Melting Pt: ∼ 82 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00661903
CAS Number: 34761-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR