6-Methoxyanthranilic acid 97%

Supplier: Thermo Fisher Scientific
Warning

H34128.03
H34128.03EA 56 EUR
H34128.03
6-Methoxyanthranilic acid 97%
6-Methoxyanthranilic acid

Purity: 97% CAS No.: 53600-33-2 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Melting Point: ca 85° dec. MDL No.: MFCD01941328

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: ∼ 85 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01941328
CAS Number: 53600-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR