3-Amino-4-chloro-1H-indazole 97%

Supplier: Thermo Fisher Scientific
Warning

H34143.MD H34143.03 H34143.06
H34143.MDEA 53 EUR
H34143.MD H34143.03 H34143.06
3-Amino-4-chloro-1H-indazole 97%
3-Amino-4-chloro-1H-indazole
Formula: C₇H₆ClN₃
Melting Pt: 157…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00663139
CAS Number: 20925-60-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR