trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: trans-4-(4-Chlorophenyl)cyclohexanecarboxylic acid

H34384.06 H34384.14 H34384.22
H34384.06EA 0 EUR
H34384.06 H34384.14 H34384.22
trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid 98%
trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid

Purity: 98% CAS No.: 49708-81-8 Molecular Formula: C13H15ClO2 Formula Weight: 238.71 Melting Point: 249-254° MDL No.: MFCD06797626

Formula: C₁₃H₁₅ClO₂
MW: 238.71 g/mol
Melting Pt: 249…254 °C
Storage Temperature: Ambient
MDL Number: MFCD06797626
CAS Number: 49708-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR