1-Eicosanethiol 98%

Supplier: Thermo Fisher Scientific

H34498.MD H34498.03
H34498.MDEA 62.6 EUR
H34498.MD H34498.03
1-Eicosanethiol 98%
1-Eicosanethiol

Purity: 98% CAS No.: 13373-97-2 Molecular Formula: C20H42S Formula Weight: 314.61 Storage Temperature: 2°C to 8°C Odour: Stench Melting Point: 38-41° MDL No.: MFCD07779406

Formula: C₂₀H₄₂S
Melting Pt: 38…41 °C
MDL Number: MFCD07779406
CAS Number: 13373-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR