Methyl imidazo[1,2-a]pyridine-7-carboxylate 95%

Supplier: Thermo Scientific
Warning

H35310.03
H35310.03EA 71 EUR
H35310.03
Methyl imidazo[1,2-a]pyridine-7-carboxylate 95%
Methyl imidazo[1,2-a]pyridine-7-carboxylate
Formula: C₉H₈N₂O₂
MW: 176,18 g/mol
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD09910350
CAS Number: 86718-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR