Ethyl imidazo[1,2-a]pyridine-2-carboxylate 95%

Supplier: Thermo Fisher Scientific
Warning

H35384.09
H35384.09EA 0 EUR
H35384.09
Ethyl imidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl imidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₀H₁₀N₂O₂
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD02325394
CAS Number: 38922-77-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR