2-Amino-3-bromo-5-fluoropyridine 97%

Supplier: Thermo Fisher Scientific
Danger

H37257.06
H37257.06EA 119 EUR
H37257.06
2-Amino-3-bromo-5-fluoropyridine 97%
2-Amino-3-bromo-5-fluoropyridine
Formula: C₅H₄BrFN₂
MW: 191 g/mol
Boiling Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD03092924
CAS Number: 869557-43-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR