3-Bromo-1H-indazole ≥97%

Supplier: Thermo Fisher Scientific
Danger

H37775.03 H37775.09
H37775.03EA 60.6 EUR
H37775.03 H37775.09
3-Bromo-1H-indazole ≥97%
3-Bromo-1H-indazole
Formula: C₇H₅BrN₂
Melting Pt: 148…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00159926
CAS Number: 40598-94-5
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR