2-Isopropoxypyridine-3-carboxaldehyde 98%

Supplier: Thermo Fisher Scientific
Warning

H50089.03 H50089.06
H50089.03EA 122 EUR
H50089.03 H50089.06
2-Isopropoxypyridine-3-carboxaldehyde 98%
2-Isopropoxypyridine-3-carboxaldehyde

Purity: 98% CAS No.: 885278-10-4 Molecular Formula: C9H11NO2 Formula Weight: 165.19 MDL No.: MFCD06804560

Formula: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD06804560
CAS Number: 885278-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR