5-(4-Nitrophenyl)-2-furoyl chloride

Supplier: Thermo Fisher Scientific

H50408.MDEA 0 EUR
H50408.MD
5-(4-Nitrophenyl)-2-furoyl chloride
5-(4-Nitrophenyl)-2-furoyl chloride

CAS No.: 60335-99-1 Molecular Formula: C11H6ClNO4 Formula Weight: 251.63 UN No.: UN3261 MDL No.: MFCD02258038

Formula: C₁₁H₆ClNO₄
MDL Number: MFCD02258038
CAS Number: 60335-99-1
UN: 3261
ADR: ,

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR