4-[4-(Trifluoromethoxy)phenoxy]piperidine ≥97%

Supplier: Thermo Fisher Scientific
Warning

H50812.03
H50812.03EA 0 EUR
H50812.03
4-[4-(Trifluoromethoxy)phenoxy]piperidine ≥97%
4-[4-(Trifluoromethoxy)phenoxy]piperidine

Purity: 97% CAS No.: 287952-67-4 Molecular Formula: C12H14F3NO2 Formula Weight: 261.25 Melting Point: 71-73° MDL No.: MFCD06656166

Formula: C₁₂H₁₄F₃NO₂
Melting Pt: 71…73 °C
Storage Temperature: Ambient
MDL Number: MFCD06656166
CAS Number: 287952-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR