3-(3-Piperidinyloxy)benzonitrile ≥99%

Supplier: Thermo Fisher Scientific
Warning

H50939.03
H50939.03EA 169 EUR
H50939.03
3-(3-Piperidinyloxy)benzonitrile ≥99%
3-(3-Piperidinyloxy)benzonitrile

Purity: 99% CAS No.: 902836-93-5 Molecular Formula: C12H14N2O Formula Weight: 202.26 MDL No.: MFCD08061079

Formula: C₁₂H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD08061079
CAS Number: 902836-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR