4-Aminomethyl-1-(n-butyl)piperidine 97%

Supplier: Thermo Fisher Scientific
Warning

H50947.MDEA 0 EUR
H50947.MD
4-Aminomethyl-1-(n-butyl)piperidine 97%
4-Aminomethyl-1-(n-butyl)piperidine

Purity: 97% CAS No.: 65017-57-4 Molecular Formula: C10H22N2 Formula Weight: 170.30 MDL No.: MFCD08060490

Formula: C₁₀H₂₂N₂
Storage Temperature: Ambient
MDL Number: MFCD08060490
CAS Number: 65017-57-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR