4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester 97%

Supplier: Thermo Fisher Scientific
Warning

H51130.03
H51130.03EA 0 EUR
H51130.03
4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester 97%
4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester

Molecular Formula: C18H22BNO4 Formula Weight: 327.18 MDL No.: MFCD11113044

Formula: C₁₈H₂₂BNO₄
Storage Temperature: Ambient
MDL Number: MFCD11113044
CAS Number: 1218790-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR