3-Methoxypiperidine hydrochloride

Supplier: Thermo Fisher Scientific
Warning

H51138.03
H51138.03EA 0 EUR
H51138.03
3-Methoxypiperidine hydrochloride
3-Methoxypiperidine hydrochloride

CAS No.: 688809-94-1 Molecular Formula: C6H13NO.HCl Formula Weight: 151.64 MDL No.: MFCD06800959

Formula: C₆H₁₃NO·HCl
MW: 151.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06800959
CAS Number: 688809-94-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR