4,5-Diphenyl-1,3-thiazol-2-amine 97%

Supplier: Thermo Fisher Scientific
Warning

H51818.03 H51818.06
H51818.03EA 0 EUR
H51818.03 H51818.06
4,5-Diphenyl-1,3-thiazol-2-amine 97%
4,5-Diphenyl-1,3-thiazol-2-amine

Purity: 97% CAS No.: 6318-74-7 Molecular Formula: C15H12N2S Formula Weight: 252.33 Odour: Odourless MDL No.: MFCD00438803

Formula: C₁₅H₁₂N₂S
Storage Temperature: Ambient
MDL Number: MFCD00438803
CAS Number: 6318-74-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR