TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 0.5 M in water 95%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3,3',3''-Phosphinetriyltripropanoic acid hydrochloride, TCEP, Tris(2-carboxyethyl)phosphine, hydrochloride

H51864.AA H51864.AC H51864.AE
H51864.AAEA 76.3 EUR
H51864.AA H51864.AC H51864.AE
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 0.5 M in water 95%
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)

Purity: 95% CAS No.: 51805-45-9 Molecular Formula: C9H15O6P.HCl Formula Weight: 286.65 UN No.: 3265 MDL No.: MFCD00145469

Formula: C₉H₁₆ClO₆P
MW: 286.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00145469
CAS Number: 51805-45-9
UN: 3265
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR