Boc-L-3,3-diphenylalanine 95%

Supplier: Thermo Fisher Scientific

H52063.06 H52063.03
H52063.06EA 430 EUR
H52063.06 H52063.03
Boc-L-3,3-diphenylalanine 95%
Boc-L-3,3-diphenylalanine

Purity: 95% CAS No.: 138662-63-2 Molecular Formula: C20H23NO4 Formula Weight: 341.41 Melting Point: 160° dec. Beilstein Reference: 4880641 MDL No.: MFCD00191186

Formula: C₂₀H₂₃NO₄
Melting Pt: 160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00191186
CAS Number: 138662-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR