Fmoc-O-t-butyl-L-β-homoserine 95%

Supplier: Thermo Fisher Scientific

H52065.03 H52065.MD
H52065.03EA 512 EUR
H52065.03 H52065.MD
Fmoc-O-t-butyl-L-β-homoserine 95%
Fmoc-O-t-butyl-L-beta-homoserine

Purity: 95% CAS No.: 203854-51-7 Molecular Formula: C23H27NO5 Formula Weight: 397.47 Melting Point: 96-98° Beilstein Reference: 8014825 MDL No.: MFCD01862865

Formula: C₂₃H₂₇NO₅
Melting Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD01862865
CAS Number: 203854-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR